Order Entry
Italy
ContactUsLinkComponent
 
 
4-Acetoxy-3-methoxybenzaldehyde 96%
 
undefined
 
4-Acetoxy-3-methoxybenzaldehyde 96%

 

  • Appearance:
    White to beige Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    73 to 80 °C
  • GC:
    ≥97.5 %

 

 

Frequently Bought Together