Order Entry
Italy
ContactUsLinkComponent
Lithium tri-tert-butoxyaluminum hydride 1.1 M in THF, AcroSeal®
Irritant
Corrosive
Flammable
Health
Lithium tri-tert-butoxyaluminum hydride 1.1 M in THF, AcroSeal®
Certificates
undefined
Irritant
Corrosive
Flammable
Health
Lithium tri-tert-butoxyaluminum hydride 1.1 M in THF, AcroSeal®

Specification Test Results

  • Appearance:
    Clear to hazy colorless Solution (May develop some turbidity or precipitate)
  • Assay:
    28 to 32 % (LTTBA)

Specification Test Results

Frequently Bought Together