



Trimethylsilylmethylmagnesium chloride 1.3 M in THF, AcroSeal®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13170-43-9
:
Formula:C₄H₁₁ClMgSi
MDL Number:MFCD00009916
:146.97 g/mol
Density:1.04 g/cm³
Flash Pt:–17 °C
UN:3399
ADR:4.3,I
:GHS07
:GHS08
:GHS05
:GHS02
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear to hazy grey to brown-black Solution
- Titration Grignard:1.2 to 1.5 M
Specification Test Results
Related Information
Frequently Bought Together