Order Entry
Italy
ContactUsLinkComponent
(Trimethylsilyl)methyllithium 10% (w/w) 0.7 M in hexanes, AcroSeal®
Flammable
Corrosive
Irritant
Health
Environment
(Trimethylsilyl)methyllithium 10% (w/w) 0.7 M in hexanes, AcroSeal®
Certificates
undefined
Flammable
Corrosive
Irritant
Health
Environment
(Trimethylsilyl)methyllithium 10% (w/w) 0.7 M in hexanes, AcroSeal®

Specification Test Results

  • Appearance:
    Colorless to yellow-brown Solution
  • Assay:
    7 to 14 % w/w

Specification Test Results

Frequently Bought Together