
4'-Trifluoromethoxyacetophenone 98%
Catalog # ACRO306230050
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:Colorless to yellow Liquid
- Infrared spectrum:Conforms
- GC:≥97.5 %
- NMR (1H-NMR):Consistent with structure
Specification Test Results
About this item
:
:204.15
Boiling Pt:110 °C (30 mmHg)
Density:1.278 g/cm³
Flash Pt:87 °C
:GHS07
Storage Temperature:Ambient
CAS Number:85013-98-5
Formula:C₉H₇F₃O₂
MDL Number:MFCD00042404