L-Lysine 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:56-87-1
:
Formula:C₆H₁₄N₂O₂
MDL Number:MFCD00064433
:146.19 g/mol
Boiling Pt:290 °C (1013 hPa)
Melting Pt:214…216 °C (dec.)
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to yellow Crystalline powder
- Infrared spectrum:Conforms
- Water:≤5.0 % (K.F.)
- Specific optical rotation:+23° to +27° (20°C, 589 nm) (c=2, 1N HCl)
- Titration with HClO4:≥97.5 % (on dry substance)
Specification Test Results
Frequently Bought Together