Inserisci ordine
Italy
ContactUsLinkComponent
2-(2-Bromoethyl)-1,3-dioxane 98%
Irritant
Health
2-(2-Bromoethyl)-1,3-dioxane 98%
Certificates
undefined
Irritant
Health
2-(2-Bromoethyl)-1,3-dioxane 98%

Specification Test Results

  • Appearance:
    Clear colorless to light brown Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4800 to 1.4840 (20°C, 589 nm)
  • GC:
    ≥97.5 %

Specification Test Results

Related Information

Frequently Bought Together