4',6'-Diamidino-2-phenylindole dihydrochloride (DAPI dihydrochloride) 98%
:28718-90-3
:
:C₁₆H₁₅N₅·2HCl
:MFCD00012681
:350.25 g/mol
:Refrigerator
- Appearance:Yellow Powder
- Infrared spectrum:Conforms
- Water:≤8 % (K.F.)
- Solubility:(1 % in water) Clear
- HPLC:≥97.5 %
- Nitrogen (N):≥18 % (Elemental analysis)
Frequently Bought Together