Order Entry
Italy
ContactUsLinkComponent
N-(tert-Butyl)hydroxylamine hydrochloride 98%
Irritant
N-(tert-Butyl)hydroxylamine hydrochloride 98%
Catalog # ACRO186910010
CAS Number:  
undefined
Irritant
N-(tert-Butyl)hydroxylamine hydrochloride 98%
Catalog # ACRO186910010
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to off-white Powder or crystals and/or chunks
  • Infrared spectrum:
    Conforms
  • Melting point:
    175 to 185 °C
  • Titration Argentometric:
    ≥97.5 %
  • HPLC:
    ≥97.5 %

Specification Test Results

About this item

 :
 :125.60 g/mol
Melting Pt:176…180 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:57497-39-9
Formula:C₄H₁₁NO
MDL Number:MFCD00012598