Order Entry
Italy
ContactUsLinkComponent
Propargyl chloride 70% (w/w) in toluene
Flammable
Corrosive
Toxic
Health
Propargyl chloride 70% (w/w) in toluene
Catalog # ACRO162701000
CAS Number:  
undefined
Flammable
Corrosive
Toxic
Health
Propargyl chloride 70% (w/w) in toluene
Catalog # ACRO162701000
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4550 to 1.4570 (20°C, 589 nm)
  • Assay:
    68 to 72 %
  • Ash:
    ≤0.5 % (Na2O)
  • pH:
    4.5 to 6.5 (4 % in water)
  • Volatile matter:
    ≤5 %

Specification Test Results

About this item

 :
Boiling Pt:60…63 °C (760 mmHg)
Density:0.963 g/cm³
Flash Pt:–15 °C
ADR:3,II
 :74.51 g/mol
UN:3286
 :GHS02
 :GHS05
 :GHS06
 :GHS08
Storage Temperature:Refrigerator
CAS Number:624-65-7
Formula:C₃H₃Cl
MDL Number:MFCD00000980