Amantadine 96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:768-94-5
:
Formula:C₁₀H₁₇N
MDL Number:MFCD00074732
:151.25 g/mol
Boiling Pt:214 °C (1013 hPa)
Melting Pt:208 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to cream Powder
- Infrared spectrum:Conforms
- Water:≤15 %
- Titration with HClO4:≥95.0 % (On dry substance)
Specification Test Results
Frequently Bought Together