Order Entry
Italy
ContactUsLinkComponent
Charcoal activated, NORIT® SA 2
undefined
Charcoal activated, NORIT® SA 2
  • Environmentally Preferable:
  • Appearance:
    Black Powder
  • Loss on drying:
    ≤10 %
  • Molasses number:
    ≤390
  • GC:
    ≥96.0 %
Frequently Bought Together