Order Entry
Italy
ContactUsLinkComponent
4,4'-Dimethoxystilbene 99%
Irritant
4,4'-Dimethoxystilbene 99%
Catalog # ACRO115660100
CAS Number:  
undefined
Irritant
4,4'-Dimethoxystilbene 99%
Catalog # ACRO115660100
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 g
  • Seq:
    3236

Specifications

Specification Test Results

  • Appearance:
    White Glistening flakes
  • Infrared spectrum:
    Conforms
  • Melting point:
    213 to 215 °C
  • GC:
    ≥98.5 %
  • Cloud point:
    22 to 28 °C (1 % in water)

Specification Test Results

About this item

 :
 :240.302 g/mol
Melting Pt:213…215 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:4705-34-4
Formula:C₁₆H₁₆O₂
MDL Number:MFCD00008414