Order Entry
Italy
ContactUsLinkComponent
2,6-Dihydroxynaphthalene 97%
Irritant
2,6-Dihydroxynaphthalene 97%
Catalog # ACRO115030050
CAS Number:  
undefined
Irritant
2,6-Dihydroxynaphthalene 97%
Catalog # ACRO115030050
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Off-white to brown Powder
  • Infrared spectrum:
    Conforms
  • HPLC:
    ≥96.0 %
  • GC:
    ≥97.5 %

Specification Test Results

About this item

 :
 :160.17 g/mol
Boiling Pt:375 °C (1013 hPa)
Melting Pt:220…224 °C
Density:1,33 g/cm³ (20 °C)
 :GHS07
Storage Temperature:Ambient
CAS Number:581-43-1
Formula:C₁₀H₈O₂
MDL Number:MFCD00004082