Order Entry
Italy
ContactUsLinkComponent
1,5-Cyclooctadiene 99+% stabilised, AcroSeal®, purified by redistillation
Irritant
Flammable
Health
Environment
1,5-Cyclooctadiene 99+% stabilised, AcroSeal®, purified by redistillation
Catalog # 111411000.
CAS Number:  
undefined
Irritant
Flammable
Health
Environment
1,5-Cyclooctadiene 99+% stabilised, AcroSeal®, purified by redistillation
Catalog # 111411000.
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, AcroSeal®
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4920 to 1.4950 (20°C, 589 nm)
  • GC:
    ≥99.0 %
  • Stabilizer:
    100 to 200 ppm octadecyl 3-(3',5'-di-tert-butyl-4'-hydroxyphenyl) propionate

Specification Test Results

About this item

 :
 :108.18 g/mol
Boiling Pt:151 °C (101.3kPa)
Melting Pt:–69.5 °C
Density:0.88 g/cm³
Flash Pt:38 °C
Storage Temperature:Ambient
UN:2520
ADR:3,III
 :GHS07
 :GHS02
 :GHS08
 :GHS09
CAS Number:111-78-4
Formula:C₈H₁₂
MDL Number:MFCD00001752