VWR® TECHNICAL Silica Wool (~ 9 microns)
24950.152
- Description:Silica Wool (~ 9 microns) TECHNICAL
- Seq:01
- Environmentally Preferable:
- Pk:50 g
Our silica wool is made from pure fused quartz fiber: amorphous, continuous, tangled, white, odorless, with no volatile component and no shot. Length, shape and arrangement of fibers give the wool its curly appearance preventing compression of the padding and improves its desired insulating properties.
- High temperature use
- Resistance to thermal shocks
- Low thermal conductivity
- High electrical resistivity
- Low dielectric constant
- Good chemical resistance