

Pivalaldehyde (trimethylacetaldehyde) 96%, AcroSeal®
Catalog # 427201000.
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle, AcroSeal®
- Pk:100 mL
- Seq:2440
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3770 to 1.3800 (20°C, 589 nm)
- GC:≥95 %
Specification Test Results
About this item
:
:86.13 g/mol
Boiling Pt:74 °C (1013 hPa)
Melting Pt:6 °C
Density:0.775 g/cm³
Flash Pt:–15 °C
UN:1989
ADR:3,II
:GHS07
:GHS02
Storage Temperature:Refrigerator
CAS Number:630-19-3
Formula:C₅H₁₀O
MDL Number:MFCD00006962