Order Entry
Italy
ContactUsLinkComponent
(2,3,6-Trifluorophenyl)acetic acid 98%
Irritant
(2,3,6-Trifluorophenyl)acetic acid 98%
Catalog # ACRO426030010
CAS Number:  
undefined
Irritant
(2,3,6-Trifluorophenyl)acetic acid 98%
Catalog # ACRO426030010
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to light yellow Crystalline powder or solid
  • Infrared spectrum:
    Conforms
  • Melting point:
    111 to 115 °C
  • HPLC:
    ≥97.5 %
  • NMR (1H-NMR):
    Consistent with structure

Specification Test Results

About this item

 :
 :190.12 g/mol
Melting Pt:111…114 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:114152-23-7
Formula:C₆H₂F₃CH₂COOH
MDL Number:MFCD00061217