Order Entry
Italy
ContactUsLinkComponent
 
 
 
 
 
Hexylmagnesium bromide 0.8 M in THF, AcroSeal®
 
undefined
 
 
 
 
Hexylmagnesium bromide 0.8 M in THF, AcroSeal®

 

  • Appearance:
    Brown Solution
  • Titration Grignard:
    0.6 to 1 M
  • Melting point:
    62 to 65 °C
  • GC:
    ≥97.5 %

 

Frequently Bought Together