Order Entry
Italy
ContactUsLinkComponent
4-Mercaptobenzoic acid 90%
Irritant
4-Mercaptobenzoic acid 90%
Certificates
undefined
Irritant
4-Mercaptobenzoic acid 90%
Certificates

Specification Test Results

  • Appearance:
    White to brown Crystalline powder or crystals
  • Infrared spectrum:
    Conforms
  • Melting point:
    215 to 224 °C
  • HPLC:
    ≥88 %

Specification Test Results

Related Information

Frequently Bought Together