Order Entry
Italy
ContactUsLinkComponent
Cyclopropylmagnesium bromide 0.5 M in THF, AcroSeal®
Irritant
Corrosive
Flammable
Health
Cyclopropylmagnesium bromide 0.5 M in THF, AcroSeal®
Catalog # 397611000.
CAS Number:  
undefined
Irritant
Corrosive
Flammable
Health
Cyclopropylmagnesium bromide 0.5 M in THF, AcroSeal®
Catalog # 397611000.
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, AcroSeal®
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Colorless to light yellow to dark yellow Solution
  • Titration Grignard:
    0.4 to 0.6 M

Specification Test Results

About this item

 :
 :145.28 g/mol
Flash Pt:–17 °C
Storage Temperature:Ambient
UN:3399
ADR:4.3,I
 :GHS07
 :GHS05
 :GHS02
 :GHS08
CAS Number:23719-80-4
Formula:C₃H₅BrMg
MDL Number:MFCD00672094