Order Entry
Italy
Orders LinkContactUsLinkComponent
Lithium peroxide ≥95%

Lithium peroxide ≥95%

10 g
Catalog # ACRO389880100
Corrosive
Oxidizing
Lithium peroxide ≥95%
Catalog # ACRO389880100
CAS Number:  12031-80-0
Lithium peroxide ≥95%
Lithium peroxide ≥95%

10 g

CAS Number:  12031-80-0
Price
75,20 €
Log in for availability
1 * 10 g
Product Details & Documents
  • CAS Number:12031-80-0
  • Formula:Li₂O₂
  • MDL Number:MFCD00016184
  • Molecular Weight:45.88 g/mol
  • Density:1.2063 g/cm³ (18 °C)
  • Storage Temperature:Ambient
  • UN:1472
  • ADR:5.1,II
  •  :Density: 1.2063 g/cm³ (18 °C)
Specifications

Specification Test Results

  • Appearance:
    White to light yellow Powder
  • Titration with KMnO4:
    ≥94 %

Specifications

  • Pk:
    10 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.