
2-Bromo-4,5-difluorobenzoic acid 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:64695-84-7
:
Formula:C₇H₃BrF₂O₂
MDL Number:MFCD04973422
:237.0000 g/mol
Melting Pt:118…122 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to yellow Crystalline powder
- Infrared spectrum:Conforms
- Melting point:118 to 122 °C
- HPLC:≥96.0 %
Specification Test Results
Frequently Bought Together