Order Entry
Italy
ContactUsLinkComponent
(5-Bromopentyl)benzene 98%
Irritant
(5-Bromopentyl)benzene 98%
Certificates
undefined
Irritant
(5-Bromopentyl)benzene 98%

Specification Test Results

  • Appearance:
    Clear colorless to yellow to light orange Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5320 to 1.5340 (20°C, 589 nm)
  • GC:
    ≥97.5 %

Specification Test Results

Related Information

Frequently Bought Together