
Milrinone 97%
ACRO397870500
:
- Pack type:Glass bottle
- Pk:50 mg
- Seq:2370
- Appearance:White to light pink Powder
- Infrared spectrum:Conforms
- HPLC:≥96.0 %
:
:211.22 g/mol
:>280 °C
:Refrigerator
:3439
:6.1,III
:GHS06
:78415-72-2
:C₁₂H₉N₃O
:MFCD00133539