
N-Methylmorpholine-N-oxide 50% (w/w) in water
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7529-22-8
:
Formula:C₅H₁₁NO₂
MDL Number:MFCD00005947
:117.15 g/mol
Melting Pt:–20 °C
Flash Pt:>100 °C
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Clear colorless to yellow Solution
- Strength:49 to 51 %
Specification Test Results
Frequently Bought Together