Allylmagnesium bromide 1 M in diethyl ether, AcroSeal®
:1730-25-2
:
:C₃H₅BrMg
:MFCD00000044
:145.28 g/mol
:0.851 g/cm³
:–40 °C
:3399
:4.3,I
:GHS07
:GHS05
:GHS02
:Ambient
- Appearance:Yellow-green to grey Solution
- Titration Grignard:0.9 to 1.1 M
Frequently Bought Together