

Neopentyl bromide 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:630-17-1
:
Formula:C₅H₁₁Br
MDL Number:MFCD00000209
:151.05 g/mol
Boiling Pt:105…106 °C (767 mmHg)
Density:1.199 g/cm³
Flash Pt:7 °C
Storage Temperature:Ambient
UN:1993
ADR:3,II
:GHS07
:GHS02
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4350 to 1.4390 (20°C, 589 nm)
- GC:≥97.5 %
Specification Test Results
Frequently Bought Together