Order Entry
Italy
ContactUsLinkComponent
 
Flammable
Corrosive
Irritant
Isopropylmagnesium bromide 3 M in 2-MeTHF (2-methyltetrahydrofuran), AcroSeal®
Catalog # 426781000.
CAS Number:  
undefined
Flammable
Corrosive
Irritant
Isopropylmagnesium bromide 3 M in 2-MeTHF (2-methyltetrahydrofuran), AcroSeal®
Catalog # 426781000.
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, AcroSeal®
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Clear dark grey to dark brown Solution
  • Titration Grignard:
    2.85 to 3.20
  • GC:
    ≥99.6 % (SOCl2)
  • Color scale:
    ≤50 APHA
  • Impurity:
    ≤0.3 % (SO2)

Specification Test Results

About this item

 :
 :147.30 g/mol
Density:1.08 g/cm³
Flash Pt:–11 °C
Storage Temperature:Ambient
UN:3399
ADR:4.3,I
 :GHS02
 :GHS05
 :GHS07
CAS Number:920-39-8
Formula:C₃H₇BrMg
MDL Number:MFCD00672110