4-Hydroxy-6-methylcoumarin 98+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13252-83-0
:
Formula:C₁₀H₈O₃
MDL Number:MFCD00085738
:176.17 g/mol
Melting Pt:261…264 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Powder
- Infrared spectrum:Conforms
- Melting point:258 to 262 °C
- Titration with TBAH:≥98.0 %
Specification Test Results
Frequently Bought Together