Order Entry
Italy
ContactUsLinkComponent
1,4-Dioxaspiro[4.5]decan-8-ol 98%
1,4-Dioxaspiro[4.5]decan-8-ol 98%
Certificates
undefined
1,4-Dioxaspiro[4.5]decan-8-ol 98%

Specification Test Results

  • Appearance:
    Clear colorless Viscous liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4850 to 1.4870 (20°C, 589 nm)
  • GC:
    ≥97.5 %

Specification Test Results

Related Information

Frequently Bought Together