Ethyl-5-methyl-1H-imidazole-4-carboxylate 98%
: Ethyl 4-Methyl-1H-imidazole-5-carboxylate, 4-Methyl-1H-imidazole-5-carboxylic acid ethyl ester, Ethyl 4-methyl-5-imidazolecarboxylate, Ethyl 5-methyl-1H-imidazole-4-carboxylate
:51605-32-4
:
:C₇H₁₀N₂O₂
:MFCD00005199
:154.1665 g/mol
:204…206 °C
:Ambient
- Environmentally Preferable:
- Appearance:Off-white to tan Crystalline powder or crystals
- Infrared spectrum:Conforms
- Melting point:204 to 208 °C
- HPLC:≥97.5 %
Frequently Bought Together