



Dibutylboron trifluoromethanesulphonate 1 M in dichloromethane
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:60669-69-4
:
Formula:C₉H₁₈BF₃O₃S
MDL Number:MFCD00009669
:274.105 g/mol
Density:1.271 g/cm³
Flash Pt:27 °C
UN:2920
ADR:8,II
:GHS02
:GHS05
:GHS07
:GHS08
Storage Temperature:Freezer
Specification Test Results
- Appearance:Clear colorless to orange Solution
- Strength:0.9 to 1.1 M
Specification Test Results
Frequently Bought Together