Order Entry
Italy
ContactUsLinkComponent
 
 
(Pentamethylcyclopentadienyl)iridium(III) dichloride dimer 99%
 
undefined
 
(Pentamethylcyclopentadienyl)iridium(III) dichloride dimer 99%

 

  • Appearance:
    Orange Crystalline powder
  • Infrared spectrum:
    Conforms
  • Assay:
    ≥98.5 % (ex Ir)

 

 

Frequently Bought Together