

3-(Trifluoromethyl)styrene 97+% stabilised
Catalog # ACRO388010010
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4640 to 1.4660 (20°C, 589 nm)
- GC:≥97.0 %
- Stabilizer:About 100 ppm 4-tert-Butylcatechol
Specification Test Results
About this item
:
:172.15 g/mol
Boiling Pt:64.5 °C (40 mmHg)
Density:1.161 g/cm³
Flash Pt:42 °C
UN:1993
ADR:3,III
:GHS07
:GHS02
Storage Temperature:Refrigerator
CAS Number:402-24-4
Formula:C₉H₇F₃
MDL Number:MFCD00075481