Order Entry
Italy
ContactUsLinkComponent
 
 
Choline hydroxide 45% (w/w) in aqueous solution stabilised, pure
 
undefined
 
Choline hydroxide 45% (w/w) in aqueous solution stabilised, pure

 

  • Appearance:
    Colorless to tan Viscous solution
  • Stabilizer:
    about 0.2 % ethylenediamine
  • Titration with HClO4:
    44.0 to 48.0 %

 

 

Frequently Bought Together