Order Entry
Italy
ContactUsLinkComponent
 
 
(S)-(-)-3-(tert-Butoxycarbonyl)-4-formyl-2,2-dimethyl-1,3-oxazolidine 95%
 
undefined
 
(S)-(-)-3-(tert-Butoxycarbonyl)-4-formyl-2,2-dimethyl-1,3-oxazolidine 95%

 

  • Appearance:
    Clear colorless to yellow Viscous liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4450 to 1.4490 (20°C, 589 nm)
  • GC:
    ≥94 %
  • Specific optical rotation:
    -85° to -95° (20°C, 589 nm) (c=1, CHCl3)

 

 

Frequently Bought Together