Order Entry
Italy
ContactUsLinkComponent
Tebbe Reagent 0.5 M in toluene
Irritant
Health
Corrosive
Flammable
Tebbe Reagent 0.5 M in toluene
Catalog # ACRO368970250
CAS Number:  
undefined
Irritant
Health
Corrosive
Flammable
Tebbe Reagent 0.5 M in toluene
Catalog # ACRO368970250
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 mL
  • Seq:
    2315

Specifications

Specification Test Results

  • Appearance:
    Dark red to purple Solution
  • Strength:
    0.40 to 0.60 M

Specification Test Results

About this item

 :
 :284.62 g/mol
Density:0.927 g/cm³
Flash Pt:4 °C
Storage Temperature:Freezer
UN:2924
ADR:3,II
 :GHS07
 :GHS08
 :GHS05
 :GHS02
CAS Number:67719-69-1
Formula:C₁₃H₁₈AlClTi
MDL Number:MFCD00151575