Methyl-3-(trifluoromethyl)phenylacetate 97%
Catalog # ACRO365600050
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4500 to 1.4520 (20°C, 589 nm)
- GC:≥96.0 %
Specification Test Results
About this item
:
:218.175 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:62451-84-7
Formula:C₁₀H₉F₃O₂
MDL Number:MFCD04039762