Order Entry
Italy
ContactUsLinkComponent
2-(3-Trifluoromethylphenyl)ethylamine 98%
Corrosive
2-(3-Trifluoromethylphenyl)ethylamine 98%
Certificates
undefined
Corrosive
2-(3-Trifluoromethylphenyl)ethylamine 98%

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4635 to 1.4655 (20°C, 589 nm)
  • GC:
    ≥97.5 %

Specification Test Results

Related Information

Frequently Bought Together