
8-Hydroxyquinoline-2-carbaldehyde 98%
:14510-06-6
:
:C₁₀H₇NO₂
:MFCD00168962
:173.1681 g/mol
:96…100 °C
:GHS07
:Ambient
- Appearance:Yellow Crystalline powder
- Infrared spectrum:Conforms
- Melting point:96 to 100 °C
- GC:≥97.5 %
Frequently Bought Together