Bis(triphenylphosphine)palladium(II) diacetate 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:14588-08-0
:
Formula:C₄₀H₃₆O₄P₂Pd
MDL Number:MFCD00010013
:749.09 g/mol
Melting Pt:136 °C
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Yellow Powder
- Infrared spectrum:Conforms
- Assay:≥14 % (Pd)
Specification Test Results
Frequently Bought Together