Order Entry
Italy
ContactUsLinkComponent
Amberlite® CG-50 ion-exchange resin Type 1
Amberlite® CG-50 ion-exchange resin Type 1
Certificates
undefined
Amberlite® CG-50 ion-exchange resin Type 1
Specification Test Results
  • Appearance:
    White to off-white Powder
  • Particle size:
    ≤10 % (< 75 µm)
  • Moisture:
    ≤10 %
  • Total weight capacity:
    ≥10 meq/g