Order Entry
Italy
ContactUsLinkComponent
 
Acid Red 18 (Food Red No. 102), pure
 
undefined
Acid Red 18 (Food Red No. 102), pure
 

 

  • Appearance:
    Red to brown Powder
  • Infrared spectrum:
    Conforms
  • UV spectrum:
    Complies
  • Solubility:
    (0.1 % in water) Clear red to very dark red
  • Dye content:
    ≥75 %
  • Insoluble matter:
    ≤0.2 % (in water)

 

 

Frequently Bought Together