Poly(acrylic acid), M.W. ∼ 5,000 50% (w/w) in water
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:9003-01-4
:9062-04-8
Formula:C₃H₄O₂
MDL Number:MFCD00084394
:72.063 g/mol
Boiling Pt:100 °C (760 mmHg)
Density:1.18 g/cm³
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear to hazy liquid
- Infrared spectrum:Conforms
- Assay:48 to 52 % Solids
- Molecular weight:3000 to 5000 (approx)
Specification Test Results
Frequently Bought Together