Poly(acrylic acid), M.W. ∼ 2,000 63% (w/w) in water
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:9003-01-4
:9062-04-8
Formula:C₃H₄O₂
MDL Number:MFCD00084394
:72.063 g/mol
Boiling Pt:100 °C (760mmHg)
Density:1.23 g/cm³
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear to hazy liquid
- Infrared spectrum:Conforms
- Assay:60.0 to 65.0 % Solids
- Molecular weight:2000 (approx)
- HPLC:≥97.5 %
Specification Test Results
Related Information
Frequently Bought Together