Order Entry
Italy
ContactUsLinkComponent
4-Methyl-1,2-cyclohexanedicarboxylic anhydride (mixture of cis- and trans-isomeres) 98%
Health
Corrosive
4-Methyl-1,2-cyclohexanedicarboxylic anhydride (mixture of cis- and trans-isomeres) 98%
Certificates
undefined
Health
Corrosive
4-Methyl-1,2-cyclohexanedicarboxylic anhydride (mixture of cis- and trans-isomeres) 98%

Specification Test Results

  • Appearance:
    Clear liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4764 to 1.4784 (20°C, 589 nm)
  • GC:
    ≥97.5 %
  • Color scale:
    ≤100 APHA

Specification Test Results

Related Information

Frequently Bought Together