
1,3-Diaminoguanidine hydrochloride 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:36062-19-8
:
Formula:CH₇N₅
MDL Number:MFCD00012948
:125.56 g/mol
Melting Pt:178…182 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to almost white Powder and/or chunks
- Infrared spectrum:Conforms
- Melting point:178 to 182 °C
- Titration Argentometric:97.5 to 102.5 %
Specification Test Results
Frequently Bought Together