
Methyl-2,2,2-trichloroacetimidate 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2533-69-9
:
Formula:C₃H₄Cl₃NO
MDL Number:MFCD00001759
Boiling Pt:148…149 °C (760 mmHg)
Density:1.42 g/cm³
Flash Pt:74 °C
:176.421 g/mol
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4770 to 1.4800 (20°C, 589 nm)
- GC:≥97.5 %
Specification Test Results
Frequently Bought Together