Order Entry
Italy
ContactUsLinkComponent
Alizarin complexone dihydrate, pure indicator grade
Alizarin complexone dihydrate, pure indicator grade
Certificates
undefined
Alizarin complexone dihydrate, pure indicator grade
Certificates
Specification Test Results
  • Appearance:
    Yellow to orange to brown Powder
  • Infrared spectrum:
    Conforms
  • Water:
    ≤9.5 % (K.F.)
  • Appearance of solution:
    (0.2% soln. in 0.02 M NaOH) clear, deep red-purple
  • UV Lambda max:
    425 to 430 nm (C=10 mg/l, Methanol)
  • Titration with NaOH:
    97 to 103 % (On dry substance)
  • Absorbance:
    ≥0.42 (lambda max, after 1 hr.)(2 ml of a 0.2% soln. in 0.02 M NaOH + 5 ml acet